Assay Details
Target Gene Details
Entrez Gene ID: | 17939 |
Gene Name: | N-acetyl galactosaminidase, alpha |
Gene Aliases: |
- |
Location: |
Chr.15:82329532-82338840 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naga | NM_008669.4 | NP_032695.3 | ||
| AK077116.1 | BAC36620.1 | |||
| AK153002.1 | BAE31642.1 | |||
| CJ093426.1 |
More Information
Set Membership: |
|

Genomic Map