Assay Details
Target Gene Details
Entrez Gene ID: | 18414 |
Gene Name: | oncostatin M receptor |
Gene Aliases: |
OSMRB |
Location: |
Chr.15:6813568-6875246 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Osmr | NM_011019.3 | 12 | 1791 | NP_035149.2 |
| AB015978.1 | 12 | 2442 | BAA33725.1 | |
| AF058805.1 | 12 | 1761 | AAC40122.1 | |
| AK157508.1 | 7 | 964 | ||
| BC137703.1 | 11 | 1689 |
More Information
Set Membership: |
|

Genomic Map