Assay Details
Target Gene Details
Entrez Gene ID: | 58200 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 1A |
Gene Aliases: |
0610038N18Rik, I-1 |
Location: |
Chr.15:103530279-103538005 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r1a | NM_021391.3 | NP_067366.1 | ||
| AF281676.1 | AAG15192.1 | |||
| BC025123.1 | AAH25123.1 | |||
| CA321941.1 |
More Information
Set Membership: |
|

Genomic Map