Assay Details
Target Gene Details
Entrez Gene ID: | 58200 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 1A |
Gene Aliases: |
0610038N18Rik, I-1 |
Location: |
Chr.15:103530279-103538005 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r1a | NM_021391.3 | NP_067366.1 | ||
AF281676.1 | AAG15192.1 | |||
BC025123.1 | AAH25123.1 | |||
CA321941.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
