Assay Details
Target Gene Details
Entrez Gene ID: | 28075 |
Gene Name: | desumoylating isopeptidase 1 |
Gene Aliases: |
AI427858, AI850401, D15Wsu75e, DeSI-1, Fam152b, Pppde2 |
Location: |
Chr.15:81992523-82016221 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Desi1 | NM_134095.2 | NP_598856.1 | ||
| AK005706.1 | BAB24198.1 | |||
| AK005963.1 | BAB24342.1 | |||
| AK016347.1 | ||||
| AK030957.1 | 1 | 3075 | ||
| AK131703.1 | BAE20768.1 | |||
| AK146155.1 | BAE26939.1 | |||
| AK170731.1 | BAE41987.1 | |||
| AK189878.1 | ||||
| BC022097.1 | AAH22097.1 | |||
| BC056972.1 | AAH56972.1 | |||
| BC071205.1 | AAH71205.1 | |||
| BC131979.1 | ||||
| BC132642.1 |
Target Gene Details
Entrez Gene ID: | 14375 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 6 |
Gene Aliases: |
70kDa, G22p1, Ku70 |
Location: |
Chr.15:81987826-82040085 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrcc6 |
More Information
Set Membership: |
|

Genomic Map