Assay Details
Target Gene Details
Entrez Gene ID: | 20363 |
Gene Name: | selenoprotein P, plasma, 1 |
Gene Aliases: |
AU018766, D15Ucla1, Se-P, selp |
Location: |
Chr.15:3270767-3280508 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 20363 |
Gene Name: | selenoprotein P, plasma, 1 |
Gene Aliases: |
AU018766, D15Ucla1, Se-P, selp |
Location: |
Chr.15:3270767-3280508 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |