Assay Details
Target Gene Details
Entrez Gene ID: | 17939 |
Gene Name: | N-acetyl galactosaminidase, alpha |
Gene Aliases: |
- |
Location: |
Chr.15:82329532-82338840 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naga | NM_008669.4 | 8 | 995 | NP_032695.3 |
| AF079458.1 | 8 | 953 | AAC28851.1 | |
| AK077116.1 | 8 | 967 | BAC36620.1 | |
| AK153002.1 | 8 | 981 | BAE31642.1 | |
| BC021631.1 | 8 | 975 | AAH21631.1 |
More Information
Set Membership: |
|

Genomic Map