Assay Details
Target Gene Details
Entrez Gene ID: | 78757 |
Gene Name: | RPTOR independent companion of MTOR, complex 2 |
Gene Aliases: |
4921505C17Rik, 6030405M08Rik, AVO3, AW492497, D530039E11Rik |
Location: |
Chr.15:6708349-6800401 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rictor | NM_030168.3 | 16 | 1369 | NP_084444.3 |
| AK036149.1 | 16 | 1370 | BAC29321.2 | |
| AK082586.1 | 16 | 1360 | ||
| AY497009.1 | 16 | 1347 | AAR89074.1 | |
| AY540053.1 | 16 | 1367 | AAS46920.1 | |
| BC022906.1 | 11 | 915 | AAH22906.1 | |
| BC120903.1 | 12 | 1008 | ||
| BC120904.1 | 16 | 1367 |

Genomic Map