Assay Details
Target Gene Details
Entrez Gene ID: | 78757 |
Gene Name: | RPTOR independent companion of MTOR, complex 2 |
Gene Aliases: |
4921505C17Rik, 6030405M08Rik, AVO3, AW492497, D530039E11Rik |
Location: |
Chr.15:6708349-6800401 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rictor | NM_030168.3 | NP_084444.3 | ||
| AK019537.1 | BAB31784.1 | |||
| AK029196.1 | BAC26344.1 | |||
| AK036149.1 | BAC29321.2 | |||
| AK036492.1 | BAC29451.1 | |||
| AK054119.1 | BAC35663.1 | |||
| AK082586.1 | ||||
| AK087858.1 | BAC40027.1 | |||
| AY497009.1 | AAR89074.1 | |||
| AY540053.1 | AAS46920.1 | |||
| BC120903.1 | ||||
| BC120904.1 |

Genomic Map