Assay Details
Target Gene Details
Entrez Gene ID: | 320506 |
Gene Name: | LMBR1 domain containing 2 |
Gene Aliases: |
9930036E21Rik |
Location: |
Chr.15:9140533-9202483 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmbrd2 | NM_177178.3 | 3 | 451 | NP_796152.2 |
| AK036527.1 | 3 | 472 | ||
| AK079450.1 | 3 | 451 | BAC37649.1 | |
| AK153728.1 | 3 | 402 | BAE32158.1 | |
| BC095971.1 | 3 | 368 | AAH95971.1 | |
| BC120727.1 | 3 | 423 | ||
| BC125421.1 | 3 | 423 |
Target Gene Details
Entrez Gene ID: | 27401 |
Gene Name: | S-phase kinase-associated protein 2 (p45) |
Gene Aliases: |
4930500A04Rik, FBXL1, FWD1, p45 |
Location: |
Chr.15:9111982-9155425 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skp2 | NM_001285980.1 | NP_001272909.1 | ||
| AK006221.1 |
More Information
Set Membership: |
|

Genomic Map