Assay Details
Target Gene Details
Entrez Gene ID: | 223455 |
Gene Name: | membrane-associated ring finger (C3HC4) 6 |
Gene Aliases: |
3830408G03, F830029L24Rik, mKIAA0597 |
Location: |
Chr.15:31455894-31531048 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| March6 | NM_172606.2 | 1 | 79 | NP_766194.2 |
| AK089827.1 | 1 | 81 | BAC40971.1 | |
| AK132461.1 | 1 | 51 | ||
| BU516606.1 | 1 | 95 |
More Information
Set Membership: |
|

Genomic Map