Assay Details
Target Gene Details
Entrez Gene ID: | 83456 |
Gene Name: | Moloney leukemia virus 10-like 1 |
Gene Aliases: |
CHAMP, Csm |
Location: |
Chr.15:88982891-89055152 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 25 - Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mov10l1 | NM_031260.2 | NP_112550.2 | ||
| AF285587.1 | AAK31966.1 | |||
| AF340211.1 | AAK77049.1 | |||
| AK084786.1 | BAC39279.1 | |||
| AY303754.1 | AAP60176.1 | |||
| JF319145.1 |

Genomic Map