Assay Details
Target Gene Details
Entrez Gene ID: | 83456 |
Gene Name: | Moloney leukemia virus 10-like 1 |
Gene Aliases: |
CHAMP, Csm |
Location: |
Chr.15:88982891-89055152 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mov10l1 | NM_031260.2 | 3 | 337 | NP_112550.2 |
| AF285587.1 | 2 | 226 | AAK31966.1 | |
| AK131735.1 | 3 | 423 | ||
| BG086707.2 | 3 | 337 | ||
| JF319145.1 | 3 | 366 |

Genomic Map