Assay Details
Target Gene Details
Entrez Gene ID: | 223455 |
Gene Name: | membrane-associated ring finger (C3HC4) 6 |
Gene Aliases: |
3830408G03, F830029L24Rik, mKIAA0597 |
Location: |
Chr.15:31455894-31531048 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| March6 | NM_172606.2 | 18 | 1848 | NP_766194.2 |
| AK028353.1 | 4 | 382 | ||
| AK042980.1 | 3 | 207 | BAC31427.1 | |
| AK089827.1 | 18 | 1850 | BAC40971.1 | |
| AK129169.1 | 6 | 593 | BAC97979.1 | |
| AK132461.1 | 18 | 1821 | ||
| BC037454.1 | 4 | 402 | AAH37454.1 | |
| BC048816.1 | 5 | 488 | AAH48816.1 | |
| BC059190.1 | 4 | 376 |
More Information
Set Membership: |
|

Genomic Map