Assay Details
Target Gene Details
Entrez Gene ID: | 68537 |
Gene Name: | mitochondrial ribosomal protein L13 |
Gene Aliases: |
1110002D09Rik |
Location: |
Chr.15:55534095-55557312 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl13 | NM_026759.3 | 2 | 110 | NP_081035.1 |
| AB049641.1 | 2 | 88 | BAB40846.1 | |
| AK003283.2 | 2 | 111 | BAB22688.1 | |
| AK141731.1 | 2 | 75 | ||
| AK160744.1 | 2 | 107 | ||
| BC027542.1 | 2 | 94 | ||
| BC055470.1 | 2 | 74 | AAH55470.1 | |
| BC060968.1 | 2 | 116 | AAH60968.1 |
More Information
Set Membership: |
|

Genomic Map