Assay Details
Target Gene Details
Entrez Gene ID: | 56612 |
Gene Name: | prefoldin 5 |
Gene Aliases: |
1190001O17Rik, 1700010A06Rik, D15Ertd697e, Eig1, MM-1 |
Location: |
Chr.15:102326116-102331490 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pfdn5 | NM_027044.3 | NP_081320.2 | ||
| AF108357.1 | AAD28373.1 | |||
| AK004438.1 | ||||
| AK005682.1 | BAB24185.2 | |||
| AK028206.1 | BAC25814.1 | |||
| AK145883.1 | BAE26723.1 | |||
| AK211903.1 | ||||
| BC026920.1 | AAH26920.1 | |||
| CN835522.1 |
More Information
Set Membership: |
|

Genomic Map