Assay Details
Target Gene Details
Entrez Gene ID: | 105837 |
Gene Name: | Mdm2, transformed 3T3 cell double minute p53 binding protein |
Gene Aliases: |
AI429604, MDM2BP |
Location: |
Chr.15:55557408-55626423 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtbp | NM_001168250.1 | 16 | 2003 | NP_001161722.1 |
| NM_134092.3 | 16 | 2003 | NP_598853.3 | |
| AJ278508.1 | 16 | 1924 | CAC07970.1 | |
| AK080036.1 | 16 | 1817 | BAC37810.1 | |
| AK088549.1 | 16 | 1817 | BAC40418.1 | |
| AK088652.1 | 6 | 1431 | BAC40479.1 | |
| AK133855.1 | 16 | 1851 | BAE21889.1 | |
| BC005500.1 | 9 | 1000 | AAH05500.1 | |
| BC052197.1 | 16 | 2003 | AAH52197.1 |

Genomic Map