Assay Details
Target Gene Details
Entrez Gene ID: | 52683 |
Gene Name: | non-SMC condensin II complex, subunit H2 |
Gene Aliases: |
- |
Location: |
Chr.15:89355719-89372850 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncaph2 | NM_001115132.2 | NP_001108604.1 | ||
| NM_001271600.1 | NP_001258529.1 | |||
| NM_001271601.1 | NP_001258530.1 | |||
| NR_073376.1 | ||||
| AK002487.1 | ||||
| AK037175.1 | BAC29736.1 | |||
| AK080447.1 | BAC37919.1 | |||
| AK083981.1 | ||||
| AK085584.1 | BAC39479.1 | |||
| AK088294.1 | BAC40265.1 | |||
| AK146642.1 | BAE27325.1 | |||
| AK152158.1 | BAE30993.1 | |||
| AK153528.1 | BAE32067.1 | |||
| AK163958.1 | BAE37553.1 | |||
| AK165250.1 | BAE38104.1 | |||
| BC003900.1 | AAH03900.1 |

Genomic Map