Assay Details
Target Gene Details
Entrez Gene ID: | 54615 |
Gene Name: | neuropeptide FF-amide peptide precursor |
Gene Aliases: |
- |
Location: |
Chr.15:102523839-102524942 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npff | NM_018787.1 | 3 | 351 | NP_061257.1 |
| NR_033409.1 | 2 | 907 | ||
| AF148701.1 | 3 | 351 | AAD39829.1 | |
| BC107294.1 | 2 | 581 | AAI07295.1 | |
| BC107295.1 | 2 | 586 | AAI07296.1 |

Genomic Map