Assay Details
Target Gene Details
Entrez Gene ID: | 109270 |
Gene Name: | proline rich 5 (renal) |
Gene Aliases: |
AU043908, Arhgap8, C030017C09Rik, C78947, Protor-1, Protor1 |
Location: |
Chr.15:84663171-84706409 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prr5 | NM_146061.4 | NP_666173.4 | ||
| AF482997.1 | AAO39849.1 | |||
| AK046057.1 | BAC32586.1 | |||
| AK081231.1 | BAC38171.1 | |||
| BC024991.1 | AAH24991.1 |

Genomic Map