Assay Details
Target Gene Details
Entrez Gene ID: | 19219 |
Gene Name: | prostaglandin E receptor 4 (subtype EP4) |
Gene Aliases: |
EP4, Ptgerep4 |
Location: |
Chr.15:5233399-5244187 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptger4 | NM_001136079.2 | NP_001129551.1 | ||
| NM_008965.2 | NP_032991.1 | |||
| AK145045.1 | BAE26204.1 | |||
| AK156428.1 | ||||
| BC009023.1 | AAH09023.1 | |||
| BC011193.1 | AAH11193.1 | |||
| D13458.1 | BAA02714.1 |
More Information
Set Membership: |
|

Genomic Map