Assay Details
Target Gene Details
Entrez Gene ID: | 20442 |
Gene Name: | ST3 beta-galactoside alpha-2,3-sialyltransferase 1 |
Gene Aliases: |
5330418N22Rik, AI467004, ST3GalI, Siat4, Siat4a, St3gal-1 |
Location: |
Chr.15:67102875-67176910 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St3gal1 | NM_009177.4 | 5 | 1383 | NP_033203.1 |
| AK031344.1 | 5 | 1273 | BAC27356.1 | |
| AK078469.1 | BAC37290.1 | |||
| AK138747.1 | 5 | 1332 | BAE23767.1 | |
| BC084730.1 | 2 | 584 | AAH84730.1 | |
| BC099670.1 | 5 | 1270 | AAH99670.1 | |
| BC099693.1 | 6 | 1350 | AAH99693.1 | |
| X73523.1 | 2 | 401 | CAA51919.1 |
More Information
Set Membership: |
|

Genomic Map