Assay Details
Target Gene Details
Entrez Gene ID: | 18162 |
Gene Name: | natriuretic peptide receptor 3 |
Gene Aliases: |
ANP-C, ANPR-C, EF-2, NPR-C, lgj, stri |
Location: |
Chr.15:11839896-11907371 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npr3 | NM_001039181.1 | NP_001034270.1 | ||
| NM_001286395.1 | NP_001273324.1 | |||
| NM_008728.2 | NP_032754.2 | |||
| AA763613.1 | ||||
| AK079196.1 | ||||
| AK166535.1 | BAE38838.1 | |||
| BC055897.1 | AAH55897.1 | |||
| BQ831432.1 | ||||
| D78175.1 | BAA11241.1 |
More Information
Set Membership: |
|

Genomic Map