Assay Details
Target Gene Details
Entrez Gene ID: | 239510 |
Gene Name: | PHD finger protein 20-like 1 |
Gene Aliases: |
CGI-72, E130113K22Rik |
Location: |
Chr.15:66577517-66647043 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf20l1 | NM_001081409.1 | NP_001074878.1 | ||
| AK032638.1 | 13 | 2012 | BAC27964.1 | |
| AK040888.1 | BAC30733.1 | |||
| AK041825.1 | 3 | 1092 | ||
| AK053606.1 | BAC35444.1 | |||
| AK187831.1 | ||||
| BC061492.1 | AAH61492.1 | |||
| BC082777.1 | AAH82777.1 | |||
| BC145689.1 | 12 | 1728 |

Genomic Map