Assay Details
Target Gene Details
Entrez Gene ID: | 239510 |
Gene Name: | PHD finger protein 20-like 1 |
Gene Aliases: |
CGI-72, E130113K22Rik |
Location: |
Chr.15:66577517-66647043 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf20l1 | NM_001081409.1 | NP_001074878.1 | ||
| AK040888.1 | BAC30733.1 | |||
| AK041825.1 | ||||
| AK053606.1 | BAC35444.1 |

Genomic Map