Assay Details
Target Gene Details
Entrez Gene ID: | 239510 |
Gene Name: | PHD finger protein 20-like 1 |
Gene Aliases: |
CGI-72, E130113K22Rik |
Location: |
Chr.15:66577517-66647043 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf20l1 | NM_001081409.1 | NP_001074878.1 | ||
| AK032638.1 | 6 | 790 | BAC27964.1 | |
| AK047044.1 | BAC32946.1 | |||
| BC052212.1 | AAH52212.1 | |||
| BC061492.1 | AAH61492.1 | |||
| BC082777.1 | AAH82777.1 | |||
| BC145689.1 |

Genomic Map