Assay Details
Target Gene Details
Entrez Gene ID: | 17988 |
Gene Name: | N-myc downstream regulated gene 1 |
Gene Aliases: |
CAP43, CMT4D, DRG1, HMSNL, NMSL, Ndr1, Ndrl, PROXY1, RTP, TDD5 |
Location: |
Chr.15:66929318-66969641 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndrg1 | NM_008681.2 | NP_032707.2 | ||
| AA795988.1 | ||||
| AK004548.1 | BAB23362.1 | |||
| AK098075.1 | ||||
| AK161860.1 | BAE36611.1 | |||
| AK165007.1 | ||||
| AK170281.1 | BAE41683.1 | |||
| AK170439.1 | BAE41797.1 | |||
| AK170636.1 | BAE41927.1 | |||
| AK170809.1 | BAE42042.1 | |||
| BC015282.1 | AAH15282.1 | |||
| BC071235.1 | AAH71235.1 | |||
| BC141246.1 | ||||
| BC171968.1 | ||||
| CA320478.1 | ||||
| U52073.1 | AAB58249.1 | |||
| U60593.1 | AAB03484.1 |
More Information
Set Membership: |
|

Genomic Map