Assay Details
Target Gene Details
Entrez Gene ID: | 73826 |
Gene Name: | polymerase (DNA-directed), delta interacting protein 3 |
Gene Aliases: |
1110008P04Rik, AA408269, AL022852, C77954, PDIP46, SKAR, mKIAA1649 |
Location: |
Chr.15:83125976-83149373 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Poldip3 | NM_178627.3 | NP_848742.1 | ||
| AK081139.1 | BAC38144.1 | |||
| AK083924.1 | BAC39068.1 | |||
| AK087446.1 | BAC39877.1 | |||
| AK088088.1 | ||||
| AK150130.1 | BAE29328.1 | |||
| AK152060.1 | BAE30916.1 | |||
| AK167552.1 | BAE39617.1 | |||
| AK220221.1 | BAD90146.1 | |||
| BC037652.1 | AAH37652.2 | |||
| BC046505.1 | AAH46505.2 | |||
| BC058225.1 | AAH58225.2 |
More Information
Set Membership: |
|

Genomic Map