Assay Details
Target Gene Details
Entrez Gene ID: | 18693 |
Gene Name: | protein interacting with C kinase 1 |
Gene Aliases: |
Prkcabp |
Location: |
Chr.15:79229166-79249466 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pick1 | NM_001045558.1 | NP_001039023.1 | ||
| NM_008837.3 | NP_032863.2 | |||
| AK050224.1 | ||||
| AK162724.1 | ||||
| BC048788.1 | AAH48788.1 | |||
| BU057774.1 | ||||
| BU698586.1 | ||||
| CA468666.1 | ||||
| CK791908.1 |
More Information
Set Membership: |
|

Genomic Map