Assay Details
Target Gene Details
Entrez Gene ID: | 69833 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide F |
Gene Aliases: |
1810060D16Rik |
Location: |
Chr.15:79141367-79151767 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2f | NM_027231.1 | NP_081507.1 | ||
| AF176516.2 | ||||
| AK007913.1 | BAB25345.1 | |||
| AK165600.1 | BAE38285.1 | |||
| AK169434.1 | BAE41175.1 | |||
| BC024419.1 | AAH24419.1 |
More Information
Set Membership: |
|

Genomic Map