Assay Details
Target Gene Details
Entrez Gene ID: | 14000 |
Gene Name: | drosha, ribonuclease type III |
Gene Aliases: |
1110013A17Rik, AI874853, Etohi2, Rn3, Rnasen |
Location: |
Chr.15:12824804-12935291 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Drosha | NM_001130149.1 | 24 | 3264 | NP_001123621.1 |
| NM_026799.3 | 24 | 3260 | NP_081075.3 | |
| AF440766.1 | 7 | 605 | AAN63637.1 | |
| AF533013.1 | 20 | 1928 | AAO65849.1 | |
| AK077549.1 | 18 | 2085 | ||
| AK144147.1 | 24 | 3219 | BAE25729.1 | |
| AK148640.1 | 24 | 3183 | BAE28629.1 | |
| BC050057.1 | 10 | 982 | AAH50057.1 | |
| BC055696.1 | 13 | 1205 | AAH55696.1 | |
| BC057687.1 | 17 | 1692 | AAH57687.1 | |
| BC060265.1 | 21 | 2559 | AAH60265.1 | |
| BC088999.1 | 24 | 3198 | AAH88999.1 |
More Information
Set Membership: |
|

Genomic Map