Assay Details
Target Gene Details
Entrez Gene ID: | 223701 |
Gene Name: | MKL (megakaryoblastic leukemia)/myocardin-like 1 |
Gene Aliases: |
AMKL, Bsac, Mal, Mrtf-A |
Location: |
Chr.15:81012281-81190757 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mkl1 | NM_001082536.1 | 1 | 130 | NP_001076005.1 |
| AF532597.1 | AAN33041.1 | |||
| AK044188.1 | 1 | 123 | BAC31809.1 | |
| AK154561.1 | 1 | 110 | BAE32678.1 | |
| AK155821.1 | 1 | 132 | ||
| BC050941.1 | AAH50941.1 | |||
| BY750304.1 | 1 | 132 |
More Information
Set Membership: |
|

Genomic Map