Assay Details
Target Gene Details
Entrez Gene ID: | 83396 |
Gene Name: | GLIS family zinc finger 2 |
Gene Aliases: |
Gli5, Klf16, Nkl |
Location: |
Chr.16:4594713-4625141 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glis2 | NM_031184.3 | NP_112461.2 | ||
| AF249340.1 | AAK28410.1 | |||
| AF325913.1 | AAK00953.1 | |||
| BC021517.1 | AAH21517.1 | |||
| CF727873.1 |
More Information
Set Membership: |
|

Genomic Map