Assay Details
Target Gene Details
Entrez Gene ID: | 67838 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 11 |
Gene Aliases: |
1810031F23Rik, ABBP-2, AL024055, Dj9, ERdj3, ERj3p |
Location: |
Chr.16:22857845-22879634 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb11 | NM_001190804.1 | 9 | 1087 | NP_001177733.1 |
| NM_001190805.1 | 9 | 1087 | NP_001177734.1 | |
| NM_026400.5 | 9 | 1087 | NP_080676.3 | |
| AK008032.1 | 4 | 270 | ||
| AK010861.1 | 9 | 1055 | ||
| AK050500.1 | 9 | 1046 | BAC34293.1 | |
| AK075785.1 | 9 | 1069 | BAC35956.1 | |
| AK075953.1 | 9 | 1049 | BAC36079.1 | |
| AK144516.1 | ||||
| AK148511.1 | 9 | 1067 | BAE28593.1 | |
| AK166730.1 | 9 | 1045 | BAE38977.1 | |
| AK205471.1 | ||||
| AY054981.1 | 9 | 963 | AAL17676.1 | |
| BC003999.2 | 9 | 1018 | AAH03999.1 | |
| BC018282.1 | 9 | 1014 | AAH18282.1 | |
| BC040747.1 | 9 | 1018 | AAH40747.1 |
More Information
Set Membership: |
|

Genomic Map