Assay Details
Target Gene Details
Entrez Gene ID: | 72026 |
Gene Name: | tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase |
Gene Aliases: |
1110005N20Rik, 1600025P05Rik, AI314320, Trmt1 |
Location: |
Chr.15:85879309-85897394 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmu | NM_028063.2 | NP_082339.1 | ||
| AK005541.1 | BAB24110.1 | |||
| AK043532.1 | BAC31570.1 | |||
| AK044059.1 | BAC31758.1 | |||
| AY349617.1 | AAQ55218.1 | |||
| BC061026.1 | AAH61026.1 |
More Information
Set Membership: |
|

Genomic Map