Assay Details
Target Gene Details
Entrez Gene ID: | 80733 |
Gene Name: | carbonic anhydrase 15 |
Gene Aliases: |
AI315043, Ca15, Cals2 |
Location: |
Chr.16:17835276-17838186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car15 | NM_030558.2 | NP_085035.1 | ||
| AF231122.1 | AAK16671.1 | |||
| AK131988.1 | BAE20923.1 | |||
| BC019975.1 | AAH19975.1 |
More Information
Set Membership: |
|

Genomic Map