Assay Details
Target Gene Details
Entrez Gene ID: | 66086 |
Gene Name: | Fgfr1op N-terminal like |
Gene Aliases: |
0610037P05Rik |
Location: |
Chr.16:14299241-14317926 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fopnl | NM_025345.2 | NP_079621.1 | ||
| AK002785.1 | ||||
| AK012206.1 | BAB28098.1 | |||
| AK084341.1 | ||||
| AK146884.1 | BAE27504.1 | |||
| AK153894.1 | BAE32240.1 | |||
| BC011300.1 | AAH11300.1 |

Genomic Map