Assay Details
Target Gene Details
Entrez Gene ID: | 223499 |
Gene Name: | DDB1 and CUL4 associated factor 13 |
Gene Aliases: |
Gm83, Wdsof1 |
Location: |
Chr.15:39112874-39146856 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf13 | NM_198606.2 | 1 | 48 | NP_941008.2 |
| BC060375.1 | AAH60375.1 | |||
| BU518555.1 | ||||
| CB950393.1 | 1 | 51 |
More Information
Set Membership: |
|

Genomic Map