Assay Details
Target Gene Details
Entrez Gene ID: | 223499 |
Gene Name: | DDB1 and CUL4 associated factor 13 |
Gene Aliases: |
Gm83, Wdsof1 |
Location: |
Chr.15:39112874-39146856 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf13 | NM_198606.2 | 9 | 1128 | NP_941008.2 |
| AK139315.1 | 2 | 6063 | ||
| AK157825.1 | 2 | 4383 | ||
| BB262409.2 | 2 | 133 | ||
| BC060375.1 | 9 | 1070 | AAH60375.1 | |
| BQ572009.1 | 6 | 606 | ||
| W42174.1 |
More Information
Set Membership: |
|

Genomic Map