Assay Details
Target Gene Details
Entrez Gene ID: | 11964 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit A |
Gene Aliases: |
AI647066, Atp6a1, Atp6a2, Atp6v1a1, VA68, VPP2 |
Location: |
Chr.16:44085402-44139700 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v1a | NM_007508.5 | 12 | 1422 | NP_031534.2 |
| AK140873.1 | 12 | 1398 | BAE24506.1 | |
| AK149833.1 | 12 | 1437 | BAE29112.1 | |
| AK150742.1 | 12 | 1408 | ||
| AK152785.1 | 2 | 136 | BAE31494.1 | |
| AK153403.1 | 12 | 1423 | BAE31963.1 | |
| AK154869.1 | 12 | 1423 | BAE32890.1 | |
| AK160792.1 | 12 | 1437 | BAE36015.1 | |
| AK166857.1 | 12 | 1412 | BAE39074.1 | |
| AK170721.1 | 12 | 1423 | BAE41978.1 | |
| AK204323.1 | 2 | 94 | ||
| BC038392.1 | 12 | 1400 | AAH38392.1 |
More Information
Set Membership: |
|

Genomic Map