Assay Details
Target Gene Details
Entrez Gene ID: | 76510 |
Gene Name: | trafficking protein particle complex 9 |
Gene Aliases: |
1810044A24Rik, 2900005P22Rik, 4632408O18Rik, Ibp, Nibp, TRS130, mKIAA1882 |
Location: |
Chr.15:72589620-73061243 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc9 | NM_001164641.1 | 15 | 2413 | NP_001158113.1 |
| NM_001164642.1 | 14 | 2386 | NP_001158114.1 | |
| NM_029640.2 | 14 | 1641 | NP_083916.1 | |
| NM_180662.2 | 14 | 2386 | NP_850993.2 | |
| AK028504.1 | 14 | 2228 | ||
| AK029106.1 | 15 | 2255 | ||
| AK156739.1 | 15 | 2211 | BAE33831.1 | |
| AK173287.1 | 4 | 385 | BAD32565.1 | |
| AY630620.1 | 14 | 1639 | AAV31909.1 | |
| BC034590.1 | 14 | 1639 | AAH34590.1 | |
| BC070463.1 | 14 | 2386 | AAH70463.1 |

Genomic Map