Assay Details
Target Gene Details
Entrez Gene ID: | 11815 |
Gene Name: | apolipoprotein D |
Gene Aliases: |
- |
Location: |
Chr.16:31296192-31314808 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apod | NM_001301353.1 | NP_001288282.1 | ||
| NM_001301354.1 | NP_001288283.1 | |||
| NM_007470.4 | NP_031496.2 | |||
| AK135046.1 | BAE22397.1 | |||
| AK157917.1 | BAE34262.1 | |||
| AK157925.1 | ||||
| AK158118.1 | BAE34364.1 | |||
| AK158405.1 | BAE34491.1 | |||
| AK160729.1 | BAE35974.1 | |||
| AK162392.1 | BAE36889.1 | |||
| BC145907.1 | ||||
| BC145909.1 | ||||
| X82648.1 | CAA57974.1 |
More Information
Set Membership: |
|

Genomic Map