Assay Details
Target Gene Details
Entrez Gene ID: | 70511 |
Gene Name: | eukaryotic elongation factor 2 lysine methyltransferase |
Gene Aliases: |
5730409G15Rik, Fam86, Fam86a, eEF2-KMT |
Location: |
Chr.16:5244155-5255956 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eef2kmt | NM_027446.2 | NP_081722.1 | ||
| AK017533.1 | BAB30795.3 | |||
| AK133590.1 | BAE21738.1 | |||
| AK163493.1 | BAE37370.1 | |||
| AK198006.1 | ||||
| BC051078.1 | AAH51078.1 |

Genomic Map