Assay Details
Target Gene Details
Entrez Gene ID: | 56175 |
Gene Name: | beta-site APP-cleaving enzyme 2 |
Gene Aliases: |
1110059C24Rik, AEPLC, AI850424, ALP56, ARP1, ASP21, BAE2, CDA13, CEAP1, DRAP |
Location: |
Chr.16:97356728-97439012 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bace2 | NM_019517.4 | NP_062390.3 | ||
| AF216310.1 | AAF36599.1 | |||
| AK052309.1 | BAC34931.1 | |||
| AK078770.1 | BAC37384.1 | |||
| AK165036.1 | BAE38011.1 | |||
| AK186833.1 | ||||
| BC120773.1 | ||||
| BC131947.1 |
More Information
Set Membership: |
|

Genomic Map