Assay Details
Target Gene Details
Entrez Gene ID: | 56175 |
Gene Name: | beta-site APP-cleaving enzyme 2 |
Gene Aliases: |
1110059C24Rik, AEPLC, AI850424, ALP56, ARP1, ASP21, BAE2, CDA13, CEAP1, DRAP |
Location: |
Chr.16:97356728-97439012 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bace2 | NM_019517.4 | 7 | 1126 | NP_062390.3 |
| AF051150.1 | 3 | 275 | AAD45964.1 | |
| AF216310.1 | 7 | 1077 | AAF36599.1 | |
| AK052309.1 | 7 | 1113 | BAC34931.1 | |
| AK078770.1 | 7 | 1093 | BAC37384.1 | |
| AK165036.1 | 7 | 1062 | BAE38011.1 | |
| AK184226.1 | 4 | 366 | ||
| AK185329.1 | 4 | 366 | ||
| BC120773.1 | 7 | 1092 | ||
| BC131947.1 | 7 | 1092 |
More Information
Set Membership: |
|

Genomic Map