Assay Details
Target Gene Details
Entrez Gene ID: | 75777 |
Gene Name: | tetratricopeptide repeat domain 23-like |
Gene Aliases: |
4930401A09Rik |
Location: |
Chr.15:10499155-10559261 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc23l | NM_029430.1 | 6 | 754 | NP_083706.1 |
| AK015027.1 | 6 | 726 | BAB29682.1 | |
| BC132384.1 | 6 | 754 | ||
| BC145852.1 | 6 | 754 |

Genomic Map