Assay Details
Target Gene Details
Entrez Gene ID: | 105887 |
Gene Name: | UDP glycosyltransferases 3 family, polypeptide A1 |
Gene Aliases: |
AI746432 |
Location: |
Chr.15:9279826-9315038 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt3a1 | NM_207216.2 | NP_997099.2 | ||
| AI746748.1 | ||||
| AK128903.1 | ||||
| AK143745.1 | BAE25522.1 | |||
| BC025940.1 | AAH25940.1 | |||
| BY006031.1 |
More Information
Set Membership: |
|

Genomic Map