Assay Details
Target Gene Details
Entrez Gene ID: | 239706 |
Gene Name: | methyltransferase like 22 |
Gene Aliases: |
- |
Location: |
Chr.16:8470753-8490205 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl22 | NM_146247.1 | NP_666359.1 | ||
| AK089212.1 | ||||
| AK136614.1 | BAE23071.1 | |||
| AK203757.1 | ||||
| BC024814.1 | AAH24814.1 |
More Information
Set Membership: |
|

Genomic Map