Assay Details
Target Gene Details
Entrez Gene ID: | 223697 |
Gene Name: | Sad1 and UNC84 domain containing 2 |
Gene Aliases: |
B230369L08Rik, C030011B15, Unc84b |
Location: |
Chr.15:79724068-79742536 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sun2 | NM_001205345.1 | NP_001192274.1 | ||
| NM_001205346.1 | NP_001192275.1 | |||
| NM_194342.3 | NP_919323.2 | |||
| AK046318.1 | ||||
| AK085196.1 | 6 | 809 | ||
| AK128958.1 | BAC87662.1 | |||
| AK156246.1 | BAE33640.1 | |||
| AK171058.1 | BAE42217.1 | |||
| AK172398.1 | ||||
| AK172996.1 | BAD32274.1 | |||
| AY682987.1 | AAT90499.1 | |||
| BC098208.1 | AAH98208.1 |
More Information
Set Membership: |
|

Genomic Map