Assay Details
Target Gene Details
Entrez Gene ID: | 12469 |
Gene Name: | chaperonin containing Tcp1, subunit 8 (theta) |
Gene Aliases: |
AI132397, Cctq, Tcpq |
Location: |
Chr.16:87483325-87495869 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cct8 | NM_009840.3 | 5 | 515 | NP_033970.3 |
| AK010882.1 | 5 | 516 | BAB27244.1 | |
| AK075711.1 | 5 | 496 | ||
| AK075880.1 | 5 | 504 | BAC36025.1 | |
| AK145661.1 | 5 | 499 | BAE26573.1 | |
| AK145754.1 | 5 | 498 | BAE26629.1 | |
| AK145918.1 | 5 | 517 | BAE26749.1 | |
| AK147038.1 | 5 | 518 | BAE27627.1 | |
| AK159969.1 | 5 | 507 | ||
| AK168465.1 | 5 | 487 | ||
| AK172867.1 | 5 | 486 | BAD32145.1 | |
| AK182683.1 | 4 | 347 | ||
| AK184650.1 | 4 | 315 | ||
| AK189477.1 | 4 | 368 | ||
| BC009007.1 | 5 | 473 | AAH09007.1 | |
| Z37164.1 | 5 | 519 | CAA85521.1 |
More Information
Set Membership: |
|

Genomic Map